Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936649.1 | - | 2 | 2840 |
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 15 | 3786 |