Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009615.2 | NP_001009615.1 | 2 | 282 | ||
GenBank mRNA | DQ336111.1 | - | 2 | 282 | ||
DQ336112.1 | - | 2 | 282 | |||
DQ336113.1 | - | 2 | 282 | |||
DQ336114.1 | - | 2 | 282 | |||
DQ336115.1 | - | 2 | 282 |