Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277353.1 | NP_001264282.1 | 10 | 1217 | ||
XM_011531650.2 | XP_011529952.1 | 10 | 2639 | |||
XM_011531651.2 | XP_011529953.1 | 10 | 1390 | |||
XM_011531652.2 | XP_011529954.1 | 11 | 1315 | |||
XM_011531653.2 | XP_011529955.1 | 11 | 1247 | |||
XM_011531654.2 | XP_011529956.1 | 10 | 2612 | |||
XM_011531656.2 | XP_011529958.1 | 9 | 2531 | |||
XM_011531657.2 | XP_011529959.1 | 10 | 1018 | |||
XM_011531658.2 | XP_011529960.1 | 8 | 2372 | |||
XM_011531663.2 | XP_011529965.1 | 8 | 2391 | |||
XM_011531664.2 | XP_011529966.1 | 7 | 952 | |||
XM_011531665.2 | XP_011529967.1 | 7 | 2232 | |||
GenBank mRNA | BC041818.1 | - | 7 | 3718 | ||
BI826002.1 | - | 4 | 568 |