Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037225.2 | NP_001032302.1 | 9 | 777 | ||
NM_001300803.1 | NP_001287732.1 | 7 | 607 | |||
NM_001318808.1 | NP_001305737.1 | 10 | 1555 | |||
XM_005273918.2 | XP_005273975.1 | 10 | 835 | |||
XM_006718502.2 | XP_006718565.1 | 10 | 952 | |||
XM_006718503.3 | XP_006718566.1 | 10 | 838 | |||
XM_006718504.2 | XP_006718567.1 | 9 | 888 | |||
XM_006718505.2 | XP_006718568.1 | 7 | 718 | |||
XM_011544940.2 | XP_011543242.1 | 10 | 833 | |||
XM_011544941.2 | XP_011543243.1 | 4 | 534 | |||
GenBank mRNA | AK093130.1 | - | 10 | 1549 | ||
BC033501.1 | - | 7 | 539 | |||
BC106951.2 | - | 8 | 572 | 40035116 | ||
BC106952.1 | - | 8 | 572 | 40035117 | ||
LC068587.1 | - | 8 | 679 |