Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032726.2 | NP_001027898.2 | 8 | 955 | ||
XM_011530948.2 | XP_011529250.1 | 7 | 2410 | |||
XM_017029488.1 | XP_016884977.1 | 8 | 931 |