Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170726.1 | NP_001164197.1 | 7 | 1024 | ||
NM_001170880.1 | NP_001164351.1 | 5 | 1237 | |||
NM_001170881.1 | NP_001164352.1 | 5 | 1255 | |||
NM_001177358.1 | NP_001170829.1 | 6 | 1289 | |||
NM_020155.3 | NP_064540.3 | 5 | 1237 | |||
XM_005274100.2 | XP_005274157.1 | 6 | 1252 | |||
XM_005274101.2 | XP_005274158.1 | 6 | 1452 | |||
XM_005274102.2 | XP_005274159.1 | 6 | 968 | |||
XM_005274104.2 | XP_005274161.1 | 6 | 916 | |||
XM_011545168.2 | XP_011543470.1 | 6 | 1162 | |||
XM_011545169.1 | XP_011543471.1 | 6 | 881 | |||
XM_011545170.2 | XP_011543472.1 | 6 | 1162 | |||
XM_011545171.2 | XP_011543473.1 | 5 | 1012 | |||
XM_011545172.2 | XP_011543474.1 | 6 | 1162 | |||
XM_017018014.1 | XP_016873503.1 | 6 | 1452 | |||
XM_017018015.1 | XP_016873504.1 | 6 | 881 | |||
XM_017018016.1 | XP_016873505.1 | 5 | 1012 | |||
GenBank mRNA | AK056094.1 | - | 6 | 1130 | ||
AK300209.1 | - | 7 | 1024 | |||
AK302256.1 | - | 6 | 1289 | |||
BC033920.1 | - | 5 | 929 | 5019895 | ||
BC035002.1 | - | 5 | 815 | 5265415 |