Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035223.3 | NP_001030300.3 | 15 | 1656 | ||
NM_001161616.2 | NP_001155088.2 | 15 | 1656 | |||
GenBank mRNA | AK057147.1 | - | 16 | 1811 | ||
AK096811.1 | - | 9 | 1261 | |||
BC014426.1 | - | 15 | 1642 | |||
BC101756.1 | - | 15 | 1642 | 8069262 | ||
BC111958.1 | - | 15 | 1642 | 8327426 | ||
BC143530.1 | - | 15 | 1642 | 9052039 |