Gene Symbol: RFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002917.1 | NP_002908.1 | 8 | 1808 | ||
XM_011523587.2 | XP_011521889.1 | 7 | 1502 | |||
GenBank mRNA | AF038196.1 | - | 5 | 1382 | ||
AK094016.1 | - | 5 | 2023 | |||
BC014495.1 | - | 4 | 1256 | |||
BC069034.1 | - | 4 | 1735 | |||
BC080181.1 | - | 1 | 710 | |||
BC146805.1 | - | 4 | 3087 | 8860343 |