Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102418.1 | NP_001095888.1 | 1295 | |||
NM_012741.1 | NP_036873.1 | 10 | 1196 | |||
GenBank mRNA | BC088155.1 | - | 1295 | 7369450 | ||
L29428.1 | - | 10 | 1196 | |||
M11884.1 | - | 850 |