Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013950.1 | NP_001013972.1 | 1 | 138 | ||
XM_008765698.1 | XP_008763920.1 | 1 | 185 | |||
XM_008765699.1 | XP_008763921.1 | 1 | 181 | |||
XM_008765700.1 | XP_008763922.1 | 1 | 189 | |||
GenBank mRNA | BC089929.1 | - | 1 | 138 | 7308748 |