Search Thermo Fisher Scientific
Gene Symbol: Msl3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014032.1 | NP_001014054.1 | 1 | 1262 | ||
XM_006256504.2 | XP_006256566.1 | 3 | 2844 | |||
GenBank mRNA | BC079056.1 | - | 1 | 1262 | 7115468 |