Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034913.1 | NP_001030085.1 | 7 | 1107 | ||
GenBank mRNA | BC105913.1 | - | 7 | 1107 | 7385857 |