Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 5 | 882 | ||
NM_001159739.2 | NP_001153211.1 | 6 | 852 | |||
NM_001287025.1 | NP_001273954.1 | 5 | 744 | |||
GenBank mRNA | DY573045.1 | - | 4 | 577 | ||
FQ210706.1 | - | 5 | 711 | |||
FQ218526.1 | - | 6 | 725 | |||
FQ218726.1 | - | 6 | 719 | |||
FQ219557.1 | - | 6 | 719 | |||
FQ219615.1 | - | 6 | 852 | |||
FQ219704.1 | - | 6 | 719 | |||
FQ231428.1 | - | 6 | 724 | |||
S72506.1 | - | 5 | 730 | |||
S82820.1 | - | 5 | 882 | |||
X78847.1 | - | 5 | 730 |