Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025715.1 | NP_001020886.1 | 5 | 393 | ||
XM_006247621.2 | XP_006247683.1 | 5 | 517 | |||
XM_006247622.2 | XP_006247684.1 | 5 | 485 | |||
GenBank mRNA | BC098855.1 | - | 5 | 393 | 7379878 | |
FQ226340.1 | - | 4 | 278 | |||
FQ231491.1 | - | 4 | 384 | |||
FQ234448.1 | - | 2 | 65 |