Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025718.1 | NP_001020889.1 | 9 | 1993 | ||
XM_006253451.2 | XP_006253513.1 | 10 | 2258 | |||
XM_008771397.1 | XP_008769619.1 | 11 | 2261 | |||
XM_008771398.1 | XP_008769620.1 | 11 | 2340 | |||
XM_008771399.1 | XP_008769621.1 | 11 | 2168 | |||
XM_008771400.1 | XP_008769622.1 | 10 | 2294 | |||
GenBank mRNA | BC098789.1 | - | 9 | 1993 | 7375514 |