Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240075.2 | XP_006240137.1 | 2, 3 | 682 | ||
XM_006240076.2 | XP_006240138.1 | 2, 3 | 681 | |||
XM_006240077.2 | XP_006240139.1 | 2, 3 | 670 | |||
GenBank mRNA | AY026526.1 | - | 2, 3 | 609 | ||
DQ473607.1 | - | 2, 3 | 564 |