Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019592.6 | NP_062538.5 | 16 | 2368 | ||
XM_011518862.1 | XP_011517164.1 | 16 | 2645 | |||
GenBank mRNA | AF265230.1 | - | 16 | 2366 | ||
AK002051.1 | - | 4 | 468 | |||
AK022300.1 | - | 11 | 1640 | |||
AK022532.1 | - | 16 | 2364 | |||
AK314401.1 | - | 16 | 2323 | |||
AL832910.1 | - | 2 | 207 | |||
AL834272.1 | - | 6 | 920 | |||
BC110584.1 | - | 16 | 2361 | 40009044 | ||
BC110585.1 | - | 16 | 2361 | 40009047 | ||
BC152309.1 | - | 16 | 2361 | 40009034 |