Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033063.1 | NP_001028235.1 | 3 | 953 | ||
XM_006254267.2 | XP_006254329.1 | 2 | 1412 | |||
XM_006254268.2 | XP_006254330.1 | 2 | 932 | |||
GenBank mRNA | BC100623.1 | - | 3 | 953 | 7929545 |