Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
281553
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
2
115
GenBank mRNA
DQ273679.1
-
2
115
FQ215069.1
-
179
FQ216531.1
-
179
FQ216869.1
-
179
FQ216875.1
-
209
FQ216956.1
-
192
FQ217047.1
-
179
FQ217054.1
-
179
FQ217106.1
-
208
FQ217137.1
-
179
FQ217169.1
-
179
FQ217172.1
-
179
FQ217195.1
-
179
FQ217299.1
-
179
FQ217371.1
-
179
FQ217378.1
-
179
FQ217706.1
-
179
FQ217788.1
-
179
FQ217840.1
-
179
FQ217842.1
-
179
FQ217896.1
-
178
FQ217902.1
-
179
FQ217974.1
-
208
FQ217984.1
-
179
FQ218010.1
-
179
FQ218048.1
-
209
FQ223543.1
-
179
FQ223572.1
-
179
FQ223583.1
-
179
FQ223607.1
-
179
FQ223623.1
-
179
FQ223636.1
-
179
FQ223643.1
-
179
FQ223690.1
-
179
FQ223692.1
-
179
FQ223711.1
-
179
FQ223713.1
-
179
FQ223719.1
-
179
FQ223773.1
-
191
FQ223816.1
-
200
FQ223825.1
-
179
FQ223832.1
-
179
FQ223835.1
-
209
FQ223976.1
-
179
FQ223986.1
-
179
FQ224079.1
-
179
FQ224114.1
-
208
FQ224128.1
-
179
FQ224223.1
-
179
FQ224258.1
-
179
FQ224340.1
-
179
FQ224376.1
-
194
FQ224378.1
-
179
FQ224387.1
-
179
FQ224450.1
-
179
FQ224462.1
-
179
FQ224599.1
-
179
FQ224648.1
-
179
FQ224672.1
-
179
FQ224735.1
-
180
FQ224749.1
-
179
FQ224755.1
-
208
FQ224795.1
-
179
FQ224811.1
-
179
FQ224822.1
-
179
FQ224827.1
-
178
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0