Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033890.1 | NP_001029062.1 | 4 | 401 | ||
GenBank mRNA | BC101880.1 | - | 4 | 401 | 7929941 |