Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033927.1 | NP_001029099.1 | 3 | 280 | ||
GenBank mRNA | BC127457.1 | - | 3 | 270 | 8363318 | |
CB608471.1 | - | 3 | 287 | |||
CK478110.1 | - | 3 | 280 | |||
U21721.1 | - | 3 | 283 |