Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034131.1 | NP_001029303.1 | 10 | 1440 | ||
XM_006236954.2 | XP_006237016.1 | 9 | 1166 | |||
XM_006236955.2 | XP_006237017.1 | 9 | 1681 | |||
XM_006236956.1 | XP_006237018.1 | 9 | 1103 | |||
XM_008763148.1 | XP_008761370.1 | 12 | 2182 | |||
XM_008763149.1 | XP_008761371.1 | 12 | 1584 | |||
XM_008763150.1 | XP_008761372.1 | 13 | 1659 | |||
XM_008763151.1 | XP_008761373.1 | 12 | 1776 | |||
XM_008763152.1 | XP_008761374.1 | 11 | 1762 | |||
XM_008763153.1 | XP_008761375.1 | 11 | 2308 | |||
XM_008763154.1 | XP_008761376.1 | 12 | 2179 | |||
XM_008763155.1 | XP_008761377.1 | 12 | 2180 | |||
XM_008763156.1 | XP_008761378.1 | 12 | 2183 | |||
XM_008763157.1 | XP_008761379.1 | 12 | 1581 | |||
XM_008763158.1 | XP_008761380.1 | 12 | 1584 | |||
GenBank mRNA | BC100267.1 | - | 10 | 1440 | 7383433 |