Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020121.3 | NP_064506.3 | 5 | 686 | ||
XM_011521094.1 | XP_011519396.1 | 5 | 641 | |||
XM_011521095.1 | XP_011519397.1 | 5 | 641 | |||
XM_011521096.1 | XP_011519398.1 | 5 | 641 | |||
XM_011521097.1 | XP_011519399.1 | 5 | 641 | |||
XM_011521100.2 | XP_011519402.1 | 5 | 641 | |||
XM_017020662.1 | XP_016876151.1 | 5 | 641 | |||
XM_017020663.1 | XP_016876152.1 | 5 | 605 | |||
GenBank mRNA | AF227906.2 | - | 5 | 587 | ||
AK001735.1 | - | 5 | 611 | |||
AL133051.1 | - | 2 | 66 | |||
BC017759.1 | - | 5 | 590 | |||
BC032302.1 | - | 5 | 686 | |||
BC067807.1 | - | 5 | 597 | |||
BC111021.1 | - | 5 | 604 | |||
BC125233.1 | - | 5 | 542 | 40124676 | ||
BC125234.1 | - | 5 | 542 |