Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031654.1 | NP_001026824.1 | 11 | 872 | ||
XM_006252627.2 | XP_006252689.1 | 11 | 892 | |||
GenBank mRNA | BC099106.1 | - | 11 | 872 | 7460531 |