Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cpt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034925.2 | NP_001030097.2 | 11 | 1379 | ||
XM_006229066.2 | XP_006229128.1 | 11 | 2401 | |||
XM_008759376.1 | XP_008757598.1 | 10 | 2545 | |||
XM_008759377.1 | XP_008757599.1 | 9 | 1382 | |||
GenBank mRNA | BC105882.1 | - | 2 | 142 | ||
CK598550.1 | - | 6 | 582 |