Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749128.3 | XP_003749176.2 | 2114 |
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101929.1 | - | 9 | 1472 | 7939237 |