Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035002.1 | NP_001030174.1 | 10 | 2288 | ||
XM_008760330.1 | XP_008758552.1 | 10 | 2373 | |||
GenBank mRNA | BC094514.1 | - | 10 | 2288 | 7307334 | |
EF688601.1 | - | 9 | 2101 | |||
FQ226030.1 | - | 1 | 742 |