Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029921.1 | NP_001025092.1 | 23 | 2396 | ||
XM_006252802.2 | XP_006252864.1 | 24 | 2688 | |||
XM_006252804.2 | XP_006252866.1 | 23 | 2638 | |||
GenBank mRNA | BC098004.1 | - | 23 | 2396 | 7453598 |