Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031660.1 | NP_001026830.1 | 5 | 678 | ||
GenBank mRNA | BC098953.1 | - | 5 | 678 | 7374871 |