Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Endog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034938.1 | NP_001030110.1 | 3 | 716 | ||
GenBank mRNA | AB221075.1 | - | 3 | 716 | ||
FQ232745.1 | - | 3 | 766 |