Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029923.1 | NP_001025094.1 | 10 | 1328 | ||
GenBank mRNA | BC098048.1 | - | 10 | 1328 | 7457181 |