Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020198.2 | NP_064583.2 | 2 | 422 | ||
XM_005257527.2 | XP_005257584.1 | 2 | 290 | |||
GenBank mRNA | AF226054.1 | - | 2 | 270 | ||
AK027786.1 | - | 2 | 264 | |||
AK027844.1 | - | 2 | 251 | |||
AK075387.1 | - | 3 | 351 | |||
AK225813.1 | - | 2 | 292 | |||
AK314328.1 | - | 2 | 277 | |||
BC008905.2 | - | 2 | 244 | 3688068 | ||
DB222124.1 | - | 2 | 422 |