Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034003.1 | NP_001029175.1 | 8 | 1949 | ||
XM_006235215.2 | XP_006235277.1 | 9 | 2083 | |||
XM_006235217.2 | XP_006235279.1 | 9 | 1966 | |||
XM_008762362.1 | XP_008760584.1 | 9 | 2497 | |||
GenBank mRNA | BC090021.1 | - | 4 | 1433 | ||
BC105824.1 | - | 8 | 1949 | 7372968 |