Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034003.1 | NP_001029175.1 | 7 | 976 | ||
XM_006235215.2 | XP_006235277.1 | 8 | 1110 | |||
XM_006235217.2 | XP_006235279.1 | 8 | 993 | |||
XM_008762362.1 | XP_008760584.1 | 8 | 1524 | |||
GenBank mRNA | AY325201.1 | - | 10 | 1071 | ||
BC090021.1 | - | 3 | 460 | |||
BC105824.1 | - | 7 | 976 | 7372968 |