Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033901.1 | NP_001029073.1 | 9 | 1911 | ||
GenBank mRNA | BC104695.1 | - | 9 | 1911 | 7111833 |