Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033902.2 | NP_001029074.2 | 11 | 1592 | ||
GenBank mRNA | BC103656.1 | - | 11 | 1525 | ||
BC166503.1 | - | 11 | 1592 | 7122476 |