Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031663.1 | NP_001026833.1 | 10 | 1304 | ||
XM_006237933.2 | XP_006237995.1 | 9 | 1347 | |||
GenBank mRNA | BC079473.1 | - | 10 | 1310 | ||
BC098713.1 | - | 10 | 1304 | 7322407 |