Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBX SIRNA Assays ›
Gene Symbol:
BBX
Gene Name:
BBX, HMG-box containing
Gene Aliases:
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location:
Chr.3: 107522936 - 107811329 on Build GRCh38
UniGene ID:
Hs.124366
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28241
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142568.2
NP_001136040.1
4, 5
482
NM_001276286.1
NP_001263215.1
4, 5
482
NM_020235.6
NP_064620.2
4, 5
482
XM_005247642.3
XP_005247699.1
4, 5
398
XM_005247643.3
XP_005247700.1
4, 5
410
XM_005247644.3
XP_005247701.1
5, 6
573
XM_011512999.2
XP_011511301.1
4, 5
482
XM_011513000.1
XP_011511302.1
4, 5
610
XM_011513001.1
XP_011511303.1
4, 5
450
XM_011513002.2
XP_011511304.1
4, 5
482
XM_011513003.2
XP_011511305.1
4, 5
482
XM_011513004.2
XP_011511306.1
4, 5
610
XM_011513005.2
XP_011511307.1
4, 5
482
XM_011513006.2
XP_011511308.1
4, 5
482
XM_011513007.2
XP_011511309.1
4, 5
482
XM_011513010.2
XP_011511312.1
4, 5
482
XM_017006881.1
XP_016862370.1
6, 7
623
XM_017006882.1
XP_016862371.1
5, 6
573
XM_017006883.1
XP_016862372.1
4, 5
482
GenBank mRNA
AF276948.1
-
1, 2
180
AF276949.1
-
4, 5
413
AF283775.2
-
4, 5
479
AF454941.1
-
4, 5
475
AF454942.1
-
2, 3
355
AK057010.1
-
4, 5
428
AK291757.1
-
4, 5
428
AK307668.1
-
3, 4
522
AK314434.1
-
4, 5
433
AL136769.1
-
2, 3
355
BC110903.1
-
2, 3
392
BC131718.1
-
4, 5
375
40123389
DA218136.1
-
4, 5
482
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
bone development
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline