Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033866.1 | NP_001029038.1 | 541 | |||
XM_006233850.1 | XP_006233912.1 | 552 | ||||
GenBank mRNA | BC059157.1 | - | 540 | 6918490 | ||
CO384344.1 | - | 541 | ||||
FQ219299.1 | - | 567 | ||||
FQ229667.1 | - | 516 |
Gene Symbol: LOC100912042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749433.2 | XP_003749481.1 | 4 | 552 | ||
XM_006233723.1 | XP_006233785.1 | 4 | 552 |