Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300743.1 | NP_001287672.1 | 13 | 1372 | ||
NM_020314.5 | NP_064710.4 | 15 | 1573 | |||
XM_005255435.3 | XP_005255492.1 | 15 | 1525 | |||
XM_006721065.3 | XP_006721128.1 | 13 | 1324 | |||
XM_011545896.2 | XP_011544198.1 | 15 | 1274 | |||
XM_011545897.2 | XP_011544199.1 | 8 | 641 | |||
GenBank mRNA | AF461052.2 | - | 9 | 1069 | ||
AK024693.1 | - | 11 | 843 | |||
AK095078.1 | - | 13 | 1069 | |||
AK290286.1 | - | 15 | 1270 | |||
AL136744.1 | - | 2 | 186 | |||
AL832159.1 | - | 15 | 1269 | |||
BC014900.2 | - | 14 | 1241 | 3909068 | ||
BC050464.1 | - | 15 | 1313 | 6061139 | ||
BC058845.1 | - | 15 | 1272 | 6452778 | ||
BX537867.1 | - | 15 | 1285 |