Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021300.2 | NP_067275.1 | 3, 4 | 647 | ||
GenBank mRNA | AF265350.1 | - | 3, 4 | 635 | ||
AJ972666.1 | - | 3, 4 | 647 | |||
BC172126.1 | - | 3, 4 | 594 | 9054796 |
Gene Symbol: Rhox4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039696.1 | NP_001034785.1 | 3, 4 | 690 | ||
GenBank mRNA | AJ972670.1 | - | 3, 4 | 690 |
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039698.1 | NP_001034787.1 | 3, 4 | 739 | ||
GenBank mRNA | AJ972671.1 | - | 3, 4 | 739 |