Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 14, 15 | 1973 | ||
GenBank mRNA | AK088950.1 | - | 13, 14 | 3563 | ||
AK129220.1 | - | 13, 14 | 3564 | |||
AK134073.1 | - | 2, 3 | 1219 | |||
BC026492.1 | - | 7, 8 | 918 | 4505095 | ||
BC092240.1 | - | 13, 14 | 1884 | 30662573 |