Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 2 | 417 | ||
XM_011517570.2 | XP_011515872.1 | 2 | 956 | |||
XM_017013646.1 | XP_016869135.1 | 3 | 184 | |||
XM_017013647.1 | XP_016869136.1 | 2 | 1146 | |||
GenBank mRNA | AF221594.1 | - | 2 | 374 | ||
AY044833.1 | - | 2 | 375 | |||
BC033684.1 | - | 2 | 377 | 5193403 | ||
BI756197.1 | - | 2 | 417 |