Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146438.1 | NP_666649.1 | 1 | 68 | ||
GenBank mRNA | BC125399.1 | - | 1 | 141 | 40129914 | |
BC125401.1 | - | 1 | 141 | 40129916 |