Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174100.1 | NP_001167571.1 | 7 | 718 | ||
NM_020418.3 | NP_065151.2 | 7 | 621 | |||
NM_033008.2 | NP_127501.1 | 6 | 570 | |||
NM_033010.2 | NP_127503.1 | 6 | 512 | |||
XM_005265330.2 | XP_005265387.1 | 7 | 2525 | |||
XM_005265331.2 | XP_005265388.1 | 7 | 644 | |||
XM_005265332.2 | XP_005265389.1 | 7 | 2525 | |||
XM_006713268.2 | XP_006713331.1 | 7 | 2333 | |||
XM_006713269.1 | XP_006713332.1 | 8 | 789 | |||
XM_006713270.1 | XP_006713333.1 | 6 | 561 | |||
XM_006713271.2 | XP_006713334.1 | 7 | 2525 | |||
XM_017006916.1 | XP_016862405.1 | 8 | 946 | |||
XM_017006917.1 | XP_016862406.1 | 7 | 2333 | |||
XM_017006918.1 | XP_016862407.1 | 6 | 2185 | |||
XM_017006919.1 | XP_016862408.1 | 6 | 561 | |||
XM_017006920.1 | XP_016862409.1 | 6 | 561 | |||
GenBank mRNA | AF176330.1 | - | 6 | 512 | ||
AF257770.1 | - | 4 | 984 | |||
AF257771.1 | - | 4 | 984 | |||
AK001244.1 | - | 7 | 888 | |||
AK023993.1 | - | 6 | 559 | |||
AK225815.1 | - | 7 | 696 | |||
BC003008.1 | - | 6 | 533 | 2822683 | ||
BC004153.2 | - | 6 | 533 | 2822683 | ||
BC017098.1 | - | 7 | 616 | 3508303 | ||
BX647811.1 | - | 5 | 1099 |