Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC061212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198667.1 | NP_941069.1 | 4 | 1529 | ||
GenBank mRNA | BC061212.1 | - | 4 | 1529 | 30250942 |
Gene Symbol: BC080696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080696.1 | - | 3 | 1454 |
Gene Symbol: Gm7978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270457.1 | NP_001257386.1 | 4 | 1529 |