Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Olfr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206896.2 | NP_996779.1 | 1 | 45 | ||
GenBank mRNA | BC120559.1 | - | 1 | 45 | 40129482 | |
BC120561.1 | - | 1 | 45 | 40129484 |