Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024842.1 | NP_001020013.1 | 2 | 752 | ||
GenBank mRNA | BC139233.1 | - | 2 | 798 | 8862255 | |
BC139269.1 | - | 2 | 798 | 8862291 |