Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128304.1 | NP_001121776.1 | 8 | 869 | ||
NM_001128305.1 | NP_001121777.1 | 6 | 831 | |||
NM_020353.2 | NP_065086.2 | 6 | 743 | |||
XM_005247654.2 | XP_005247711.1 | 7 | 770 | |||
XM_005247655.2 | XP_005247712.1 | 7 | 687 | |||
XM_011513031.2 | XP_011511333.1 | 6 | 629 | |||
GenBank mRNA | AF199023.1 | - | 6 | 647 | ||
AK290214.1 | - | 6 | 771 | |||
AK292815.1 | - | 8 | 769 | |||
AK312809.1 | - | 6 | 685 | |||
AL833760.1 | - | 6 | 657 | |||
AY550971.1 | - | 6 | 597 | |||
BC028354.2 | - | 6 | 743 | 4831340 |