Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011875.2 | NP_001011875.1 | 1 | 216 | ||
GenBank mRNA | BC146014.1 | - | 1 | 216 | 40131317 | |
BC146016.1 | - | 1 | 216 | 40131319 |